Please login to your online account to display your discounted pricing
Thermo Scientific™ 2.2 mL Storage Plate
2.2 mL deepwell storage plates for the culture or storage of large samples.
Manufacturer: thermo scientific™  AB0661W
 View more versions of this product
Catalog No. AB-0661W
  • / Case of 50 Each
Description & Specifications
Specifications
Type Square well
No. per Case 50
Well Design U-shaped bottom
Color White
Description Feature square well geometry which promotes efficient sample aeration, and U-bottomed wells to aid in re-suspension
Volume (Metric) Well 2.2mL
Streamline your molecular biology applications with Thermo Scientific storage and assay plates. All plates are manufactured in our cleanroom facility to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. All plates are ANSI format for compatibility with automated systems. To further assist in storage and tracking, all plates can be supplied with custom barcoding.
Highlights
  • Designed for optimal sample recovery
  • Each square well is specifically designed with an independent sealing rim to prevent cross-contamination
  • U-bottom (AB-0661) to aid resuspension or conical bottom (AB-0932) for ease of sample retrieval