missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2′-Methylenebisbenzothiazole, TCI America™

$117.64 - $330.00

Chemical Identifiers

CAS 1945-78-4
Molecular Formula C15H10N2S2
Molecular Weight (g/mol) 282.379
MDL Number MFCD00051395
InChI Key ZRXNLCFJIKBZCA-UHFFFAOYSA-N
Synonym 2,2'-methylenebisbenzothiazole, benzothiazole, 2,2'-methylenebis, bis benzo d thiazol-2-yl methane, 2-1,3-benzothiazol-2-ylmethyl-1,3-benzothiazole, 2,2'-methylenebis 1,3-benzothiazole, 2,2'-methylenedibenzothiazole, maybridge1_006808, di-2-benzothiazolylmethane, bis-2-benzothiazolylmethane, acmc-1bx67
PubChem CID 74755
IUPAC Name 2-(1,3-benzothiazol-2-ylmethyl)-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)CC3=NC4=CC=CC=C4S3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M13905G View Documents TCI America
M13905G
5g Each for $117.64
 
M139025G View Documents TCI America
M139025G
25g Each for $330.00
 
Specifications

Chemical Identifiers

1945-78-4
282.379
ZRXNLCFJIKBZCA-UHFFFAOYSA-N
74755
C1=CC=C2C(=C1)N=C(S2)CC3=NC4=CC=CC=C4S3
C15H10N2S2
MFCD00051395
2,2'-methylenebisbenzothiazole, benzothiazole, 2,2'-methylenebis, bis benzo d thiazol-2-yl methane, 2-1,3-benzothiazol-2-ylmethyl-1,3-benzothiazole, 2,2'-methylenebis 1,3-benzothiazole, 2,2'-methylenedibenzothiazole, maybridge1_006808, di-2-benzothiazolylmethane, bis-2-benzothiazolylmethane, acmc-1bx67
2-(1,3-benzothiazol-2-ylmethyl)-1,3-benzothiazole

Specifications

Yellow
5g
C15H10N2S2
2,2'-methylenebisbenzothiazole, benzothiazole, 2,2'-methylenebis, bis benzo d thiazol-2-yl methane, 2-1,3-benzothiazol-2-ylmethyl-1,3-benzothiazole, 2,2'-methylenebis 1,3-benzothiazole, 2,2'-methylenedibenzothiazole, maybridge1_006808, di-2-benzothiazolylmethane, bis-2-benzothiazolylmethane, acmc-1bx67
C1=CC=C2C(=C1)N=C(S2)CC3=NC4=CC=CC=C4S3
282.379
282.38
≥97.0% (GC)
93°C
1945-78-4
MFCD00051395
ZRXNLCFJIKBZCA-UHFFFAOYSA-N
2-(1,3-benzothiazol-2-ylmethyl)-1,3-benzothiazole
74755
Crystal-Powder at 20°C
2,2′-Methylenebisbenzothiazole
Safety and Handling

Safety and Handling

TSCA : Yes