missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2′-Methylenebis(6-tert-butyl-4-ethylphenol), TCI America™

Manufacturer:  TCI America M0215500G

 View more versions of this product

Catalog No. M0215500G



Specifications

Specifications

2,2′-Methylenebis(6-tert-butyl-4-ethylphenol)
125°C
88-24-4
MFCD00026296
GPNYZBKIGXGYNU-UHFFFAOYSA-N
2-tert-butyl-6-[(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methyl]-4-ethylphenol
6928
Crystal-Powder at 20°C
White-Yellow
500g
C25H36O2
Bis(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methane, 3,3′-Di-tert-butyl-5,5′-diethyl-2,2′-dihydroxydiphenylmethane, 2,2′-Methylenebis(4-ethyl-6-tert-butylphenol)
CCC1=CC(=C(C(=C1)C(C)(C)C)O)CC2=C(C(=CC(=C2)CC)C(C)(C)C)O
368.561
368.56
≥98.0% (GC)

Chemical Identifiers

88-24-4
368.561
GPNYZBKIGXGYNU-UHFFFAOYSA-N
6928
CCC1=CC(=C(C(=C1)C(C)(C)C)O)CC2=C(C(=CC(=C2)CC)C(C)(C)C)O
C25H36O2
MFCD00026296
Bis(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methane, 3,3′-Di-tert-butyl-5,5′-diethyl-2,2′-dihydroxydiphenylmethane, 2,2′-Methylenebis(4-ethyl-6-tert-butylphenol)
2-tert-butyl-6-[(3-tert-butyl-5-ethyl-2-hydroxyphenyl)methyl]-4-ethylphenol
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0093

RTECSNumber : SL9800000

TSCA : Yes

Documents