missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2′-Methylenebis(6-cyclohexyl-p-cresol) 97.0+%, TCI America™

$95.50 - $485.09

Chemical Identifiers

CAS 4066-02-8
Molecular Formula C27H36O2
Molecular Weight (g/mol) 392.58
InChI Key AKNMPWVTPUHKCG-UHFFFAOYSA-N
Synonym 2,2′C-Methylenebis(6-cyclohexyl-4-methylphenol), Bis(5-cyclohexyl-6-hydroxy-m-tolyl)methane
PubChem CID 77681
IUPAC Name 2-cyclohexyl-6-[(3-cyclohexyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C2CCCCC2)O)CC3=C(C(=CC(=C3)C)C4CCCCC4)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M189625G
View Documents
Tci America
M189625G
25g Each for $95.50
 
M1896250G
View Documents
Tci America
M1896250G
250g Each for $485.09
 
Specifications

Chemical Identifiers

4066-02-8
392.58
2,2′C-Methylenebis(6-cyclohexyl-4-methylphenol), Bis(5-cyclohexyl-6-hydroxy-m-tolyl)methane
2-cyclohexyl-6-[(3-cyclohexyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
C27H36O2
AKNMPWVTPUHKCG-UHFFFAOYSA-N
77681
CC1=CC(=C(C(=C1)C2CCCCC2)O)CC3=C(C(=CC(=C3)C)C4CCCCC4)O

Specifications

White-Yellow
25g
C27H36O2
AKNMPWVTPUHKCG-UHFFFAOYSA-N
2-cyclohexyl-6-[(3-cyclohexyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
77681
Crystal-Powder at 20°C
2,2′-Methylenebis(6-cyclohexyl-p-cresol)
134°C
4066-02-8
2,2′C-Methylenebis(6-cyclohexyl-4-methylphenol), Bis(5-cyclohexyl-6-hydroxy-m-tolyl)methane
CC1=CC(=C(C(=C1)C2CCCCC2)O)CC3=C(C(=CC(=C3)C)C4CCCCC4)O
392.58
392.58
≥97.0% (HPLC)
Safety and Handling

Safety and Handling

RTECSNumber : SM0335000

TSCA : Yes