missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2′-Methylenebis(6-cyclohexyl-p-cresol) 97.0+%, TCI America™

Manufacturer:  Tci America M1896250G

 View more versions of this product

Catalog No. M1896250G



Specifications

Specifications

2,2′-Methylenebis(6-cyclohexyl-p-cresol)
134°C
4066-02-8
2,2′-Methylenebis(6-cyclohexyl-4-methylphenol), Bis(5-cyclohexyl-6-hydroxy-m-tolyl)methane
CC1=CC(=C(C(=C1)C2CCCCC2)O)CC3=C(C(=CC(=C3)C)C4CCCCC4)O
392.58
392.58
≥97.0% (HPLC)
White-Yellow
250g
C27H36O2
AKNMPWVTPUHKCG-UHFFFAOYSA-N
2-cyclohexyl-6-[(3-cyclohexyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
77681
Crystal-Powder at 20°C

Chemical Identifiers

4066-02-8
392.58
2,2′-Methylenebis(6-cyclohexyl-4-methylphenol), Bis(5-cyclohexyl-6-hydroxy-m-tolyl)methane
2-cyclohexyl-6-[(3-cyclohexyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
C27H36O2
AKNMPWVTPUHKCG-UHFFFAOYSA-N
77681
CC1=CC(=C(C(=C1)C2CCCCC2)O)CC3=C(C(=CC(=C3)C)C4CCCCC4)O
Safety and Handling

Safety and Handling

RTECSNumber : SM0335000

TSCA : Yes

Documents