Please login to your online account to display your discounted pricing

2-(2-(Methylamino)ethyl)pyridine dihydrochloride, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  167990250

 View more versions of this product

Catalog No. AC167990250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 209.12
Assay ≥98.5%
Physical Form Crystalline Powder
Quantity 25g
Color White to light yellow
Beilstein 22, 434
Merck Index 15, 1180
EINECS Number 226-966-5
TSCA TSCA
Melting Point 150° to 154°C
pH 2 to 4 (2% soln.)
CAS Max % 100.0
Appearance of Solution (2% aq. soln.) clear, colorless
Sulfated Ash 0.1% max.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
CAS Min % 98.5
Infrared Spectrum Authentic
Loss on Drying 1% max. (1 g, 105°C)
Synonym βhistine dihydrochloride
Organic Impurity organic impurities: < 0.5%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula (C5H4N)CH2CH2N(H)CH3·2HCl
Name Note 99%
CAS 5579-84-0
Chemical Name or Material 2-(2-(Methylamino)ethyl)pyridine dihydrochloride
MDL Number MFCD00012813
Molecular Formula C8H12N2·2HCl
Assay Percent Range 99%

Specifications

Chemical Name or Material 2-(2-(Methylamino)ethyl)pyridine dihydrochloride
Name Note 99%
CAS 5579-84-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H12N2·2HCl
Linear Formula (C5H4N)CH2CH2N(H)CH3·2HCl
Formula Weight 209.12
MDL Number MFCD00012813
Synonym βhistine dihydrochloride
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

SDS & Safety Documents