Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine, 98%

Chemical Identifiers

Chemical Name or Material 2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine
CAS 152460-09-8
Formula Weight 307.31
MDL Number MFCD02179269
Beilstein 7544032
Synonym N-(2-Methyl-5-nitrophenyl)-4-(3-pyridyl)-2-pyrimidinylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6481403

  SDS
alfa aesar™
H6481403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6481406

  SDS
alfa aesar™
H6481406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6481414

  SDS
alfa aesar™
H6481414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 307.31
Melting Point 188° to 193°C
Chemical Name or Material 2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine
Physical Form Solid
Assay 98%
MDL Number MFCD02179269
CAS 152460-09-8
Beilstein 7544032
Synonym N-(2-Methyl-5-nitrophenyl)-4-(3-pyridyl)-2-pyrimidinylamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Yellow
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.