missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Methyl-5-nitroanilino)-4-(3-pyridyl)pyrimidine, TCI America™

$58.06 - $171.11

Chemical Identifiers

CAS 152460-09-8
Molecular Formula C16H13N5O2
Molecular Weight (g/mol) 307.313
MDL Number MFCD02179269
InChI Key OJITWRFPRCHSMX-UHFFFAOYSA-N
Synonym n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimidin-2-amine, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidine-amine, chembl20426, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidineamine, n-2-methyl-5-nitrophenyl-4-pyridin-3-ylpyrimidin-2-amine, 2-methyl-4-nitro-phenyl-4-pyridin-3-yl-pyrimidin-2-yl-amine, 2-methyl-5-nitro-n-4-pyridin-3-yl pyrimidin-2-yl benzenamine, 2-pyrimidinamine, n-2-methyl-5-nitrophenyl-4-3-pyridinyl, n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimid, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidinamine
PubChem CID 10686149
IUPAC Name N-(2-methyl-5-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])NC2=NC=CC(=N2)C3=CN=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25841G View Documents TCI America
M25841G
1g Each for $58.06
 
M25845G View Documents TCI America
M25845G
5g Each for $171.11
 
Specifications

Chemical Identifiers

152460-09-8
307.313
OJITWRFPRCHSMX-UHFFFAOYSA-N
10686149
CC1=C(C=C(C=C1)[N+](=O)[O-])NC2=NC=CC(=N2)C3=CN=CC=C3
C16H13N5O2
MFCD02179269
n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimidin-2-amine, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidine-amine, chembl20426, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidineamine, n-2-methyl-5-nitrophenyl-4-pyridin-3-ylpyrimidin-2-amine, 2-methyl-4-nitro-phenyl-4-pyridin-3-yl-pyrimidin-2-yl-amine, 2-methyl-5-nitro-n-4-pyridin-3-yl pyrimidin-2-yl benzenamine, 2-pyrimidinamine, n-2-methyl-5-nitrophenyl-4-3-pyridinyl, n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimid, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidinamine
N-(2-methyl-5-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine

Specifications

Yellow
1g
C16H13N5O2
n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimidin-2-amine, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidine-amine, chembl20426, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidineamine, n-2-methyl-5-nitrophenyl-4-pyridin-3-ylpyrimidin-2-amine, 2-methyl-4-nitro-phenyl-4-pyridin-3-yl-pyrimidin-2-yl-amine, 2-methyl-5-nitro-n-4-pyridin-3-yl pyrimidin-2-yl benzenamine, 2-pyrimidinamine, n-2-methyl-5-nitrophenyl-4-3-pyridinyl, n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimid, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidinamine
CC1=C(C=C(C=C1)[N+](=O)[O-])NC2=NC=CC(=N2)C3=CN=CC=C3
307.313
307.31
≥98.0% (HPLC,T)
194°C
152460-09-8
MFCD02179269
OJITWRFPRCHSMX-UHFFFAOYSA-N
N-(2-methyl-5-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine
10686149
Crystal-Powder at 20°C
2-(2-Methyl-5-nitroanilino)-4-(3-pyridyl)pyrimidine
Safety and Handling

Safety and Handling

TSCA : No