missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Methyl-4-thiazolyl)phenol, 97%

$79.15 - $276.74

Chemical Identifiers

CAS 160241-65-6
Molecular Formula C10H9NOS
Formula Weight 191.25
MDL Number MFCD00665949
Chemical Name or Material 2-(2-Methyl-4-thiazolyl)phenol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5043103 SDS Alfa Aesar™
H5043103
1g Each for $276.74
AAH50431MD SDS Alfa Aesar™
H50431MD
250mg Each for $79.15
Specifications Description & Specifications

Chemical Identifiers

160241-65-6
191.25
2-(2-Methyl-4-thiazolyl)phenol
C10H9NOS
MFCD00665949

Specifications

160241-65-6
191.25
61° to 64°C
2-(2-Methyl-4-thiazolyl)phenol
C10H9NOS
MFCD00665949
1g
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning