Please login to your online account to display your discounted pricing

2-(2-Methyl-1H-imidazol-1-yl)aniline, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC61214CB

 View more versions of this product

Catalog No. CC61214CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 173.22
Quantity 250mg
Assay 97%
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C10H11N3
CAS 26286-55-5
Chemical Name or Material 2-(2-Methyl-1H-imidazol-1-yl)aniline
Health Hazard 1 Warning

Specifications

Chemical Name or Material 2-(2-Methyl-1H-imidazol-1-yl)aniline
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11N3
Formula Weight 173.22
Accessories

Warning