missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(2-Methyl-1,3-thiazol-4-yl)ethylamine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC54613CB
Specifications
2-(2-Methyl-1, 3-thiazol-4-yl)ethylamine | |
97% | |
C6H10N2S | |
250mg | |
GLEZGSKBHYTSAD-UHFFFAOYSA-N | |
2-(2-methyl-1,3-thiazol-4-yl)ethanamine | |
7536518 | |
97% |
165115-15-1 | |
Amber Glass Bottle | |
MFCD03011601 | |
2-2-methyl-1,3-thiazol-4-yl ethanamine, 2-2-methylthiazol-4-yl ethanamine, 2-2-methyl-1,3-thiazol-4-yl ethylamine, 4-thiazoleethanamine, 2-methyl, 2-2-methyl-1,3-thiazol-4-yl ethan-1-amine, 4-thiazoleethanamine,2-methyl, 2-2-methyl-1,3-thiazol-4-yl-1-ethanamine | |
CC1=NC(CCN)=CS1 | |
142.22 | |
142.23 |
Chemical Identifiers
165115-15-1 | |
142.22 | |
GLEZGSKBHYTSAD-UHFFFAOYSA-N | |
7536518 | |
CC1=NC(CCN)=CS1 |
C6H10N2S | |
MFCD03011601 | |
2-2-methyl-1,3-thiazol-4-yl ethanamine, 2-2-methylthiazol-4-yl ethanamine, 2-2-methyl-1,3-thiazol-4-yl ethylamine, 4-thiazoleethanamine, 2-methyl, 2-2-methyl-1,3-thiazol-4-yl ethan-1-amine, 4-thiazoleethanamine,2-methyl, 2-2-methyl-1,3-thiazol-4-yl-1-ethanamine | |
2-(2-methyl-1,3-thiazol-4-yl)ethanamine |