Please login to your online account to display your discounted pricing

2-(2-Methyl-1,3-thiazol-4-yl)ethylamine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC54613DA

 View more versions of this product

Catalog No. CC54613DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 142.23
Quantity 1g
Assay 97%
CAS 165115-15-1
Chemical Name or Material 2-(2-Methyl-1, 3-thiazol-4-yl)ethylamine
Molecular Formula C6H10N2S
Packaging Amber Glass Bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material 2-(2-Methyl-1, 3-thiazol-4-yl)ethylamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H10N2S
Formula Weight 142.23