missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Methyl-1,3-thiazol-4-yl)aniline, 95%, Maybridge

Chemical Identifiers

CAS 305811-38-5
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C10H10N2S
Formula Weight 190.27
MDL Number MFCD09965287
Chemical Name or Material 2-(2-Methyl-1, 3-thiazol-4-yl)aniline
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC68414CB SDS Fisher Scientific Company
CC68414CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC68414DA SDS Fisher Scientific Company
CC68414DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

305811-38-5
100.0
C10H10N2S
MFCD09965287
95%
94.5
95%
190.27
2-(2-Methyl-1, 3-thiazol-4-yl)aniline

Specifications

190.27
100.0
C10H10N2S
94.5
305811-38-5
95%
250mg
95%
MFCD09965287
Amber Glass Bottle
2-(2-Methyl-1, 3-thiazol-4-yl)aniline

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning