Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[(2-Methoxyphenyl)dimethylsilyl]benzyl alcohol, 95%

$140.53 - $140.53

Chemical Identifiers

Chemical Name or Material 2-[(2-Methoxyphenyl)dimethylsilyl]benzyl alcohol
CAS 1217863-51-8
Molecular Formula C16H20O2Si
Formula Weight 272.43g/mol
MDL Number MFCD16251556
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5174206

 
alfa aesar™
H51742-06
Each for $140.53
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 2-[(2-Methoxyphenyl)dimethylsilyl]benzyl alcohol
CAS 1217863-51-8
Assay 95%
Molecular Formula C16H20O2Si
Formula Weight 272.43g/mol
MDL Number MFCD16251556
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.