missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Methoxyphenyl)benzimidazole, 95%

$124.14 - $1,628.47

Chemical Identifiers

CAS 6528-85-4
Molecular Formula C14H12N2O
Molecular Weight (g/mol) 224.263
MDL Number MFCD00447053
InChI Key JNFBDHNAZPBWTF-UHFFFAOYSA-N
Synonym 2-2-methoxyphenyl-1h-benzimidazole, 1h-benzimidazole, 2-2-methoxyphenyl, 2-2-methoxyphenyl-1h-benzo d imidazole, 2-2-methoxyphenyl-1h-1,3-benzodiazole, 2-2-methoxy-phenyl-1h-benzoimidazole, 1h-benzo d imidazole,2-2-methoxyphenyl
PubChem CID 759420
IUPAC Name 2-(2-methoxyphenyl)-1H-benzimidazole
SMILES COC1=CC=CC=C1C2=NC3=CC=CC=C3N2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64294MD View Documents Alfa Aesar™
H64294MD
250mg Each for $124.14
 
AAH6429403 View Documents Alfa Aesar™
H6429403
1g Each for $370.11
 
AAH6429406 View Documents Alfa Aesar™
H6429406
5g Each for $1,628.47
 
Specifications

Chemical Identifiers

6528-85-4
224.263
JNFBDHNAZPBWTF-UHFFFAOYSA-N
759420
COC1=CC=CC=C1C2=NC3=CC=CC=C3N2
C14H12N2O
MFCD00447053
2-2-methoxyphenyl-1h-benzimidazole, 1h-benzimidazole, 2-2-methoxyphenyl, 2-2-methoxyphenyl-1h-benzo d imidazole, 2-2-methoxyphenyl-1h-1,3-benzodiazole, 2-2-methoxy-phenyl-1h-benzoimidazole, 1h-benzo d imidazole,2-2-methoxyphenyl
2-(2-methoxyphenyl)-1H-benzimidazole

Specifications

250mg
C14H12N2O
2-2-methoxyphenyl-1h-benzimidazole, 1h-benzimidazole, 2-2-methoxyphenyl, 2-2-methoxyphenyl-1h-benzo d imidazole, 2-2-methoxyphenyl-1h-1,3-benzodiazole, 2-2-methoxy-phenyl-1h-benzoimidazole, 1h-benzo d imidazole,2-2-methoxyphenyl
COC1=CC=CC=C1C2=NC3=CC=CC=C3N2
224.263
224.26
95%
6528-85-4
MFCD00447053
JNFBDHNAZPBWTF-UHFFFAOYSA-N
2-(2-methoxyphenyl)-1H-benzimidazole
759420
Solid
2-(2-Methoxyphenyl)benzimidazole