Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Methoxyphenyl)-5-phenyl-1,3,4-oxadiazole, 98%

$107.61 - $107.61

Chemical Identifiers

Chemical Name or Material 2-(2-Methoxyphenyl)-5-phenyl-1,3,4-oxadiazole
CAS 1874-42-6
Molecular Formula C15H12N2O2
Formula Weight 252.27g/mol
MDL Number MFCD00050995
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1853406

 
alfa aesar™
A18534-06
5g Each for $107.61

AAA1853403

 
alfa aesar™
A18534-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 99° to 101°C
Quantity 5g
Chemical Name or Material 2-(2-Methoxyphenyl)-5-phenyl-1,3,4-oxadiazole
CAS 1874-42-6
Assay 98%
Molecular Formula C15H12N2O2
Formula Weight 252.27g/mol
MDL Number MFCD00050995