Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Methoxyphenoxy)-N-methylethylamine, 96%

$116.99 - $404.82

Chemical Identifiers

Chemical Name or Material 2-(2-Methoxyphenoxy)-N-methylethylamine
CAS 72955-82-9
Molecular Formula C10H15NO2
Formula Weight 181.24
MDL Number MFCD07400597
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5029903

 
alfa aesar™
H5029903
1g Each for $404.82

AAH50299MD

 
alfa aesar™
H50299MD
250mg Each for $116.99
Description & Specifications

Specifications

Formula Weight 181.24
Chemical Name or Material 2-(2-Methoxyphenoxy)-N-methylethylamine
Assay 96%
Molecular Formula C10H15NO2
MDL Number MFCD07400597
CAS 72955-82-9
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.