Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Methoxyphenoxy)ethylamine, 97%

Chemical Identifiers

Chemical Name or Material 2-(2-Methoxyphenoxy)ethylamine
CAS 1836-62-0
Molecular Formula C9H13NO2
Formula Weight 167.21
MDL Number MFCD00235185
UN Number UN2735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5025103

  SDS
alfa aesar™
H5025103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH50251MD

  SDS
alfa aesar™
H50251MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.21
Chemical Name or Material 2-(2-Methoxyphenoxy)ethylamine
Assay 97%
Molecular Formula C9H13NO2
MDL Number MFCD00235185
Boiling Point 98°C (0.4mmHg)
CAS 1836-62-0
UN Number UN2735
Quantity 1g
Odor Amine-like
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Applications
2-(2-Methoxyphenoxy)ethylamine is used for its reaction with 4-(2,3-epoxypropoxy) carbazole for preparation of Carvedilol (I).

Solubility
Difficult to mix.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.