Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Mercaptoethyl)pyrazine, 97%

Chemical Identifiers

Chemical Name or Material 2-(2-Mercaptoethyl)pyrazine
CAS 35250-53-4
Molecular Formula C6H8N2S
Formula Weight 140.21
MDL Number MFCD00053160
UN Number UN2810
Synonym 2-Pyrazinylethanethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1304503

  SDS
alfa aesar™
A1304503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1304506

  SDS
alfa aesar™
A1304506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 140.21
Chemical Name or Material 2-(2-Mercaptoethyl)pyrazine
TSCA TSCA
Assay 97%
Molecular Formula C6H8N2S
MDL Number MFCD00053160
CAS 35250-53-4
UN Number UN2810
Synonym 2-Pyrazinylethanethiol
Health Hazard 1 Danger
Boiling Point 105° to 110°C (20mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECS Number KJ2551000

Applications
2-(2-Mercaptoethyl)pyrazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals, petrochemicals and dyestuff. It is used as a flavoring agent has a sulfurous type odor and a meaty type flavor.

Solubility
Partly soluble in water, soluble in most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C.Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C