Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Mercapto-4-methyl-5-thiazolyl)ethanol, 95%

Chemical Identifiers

Chemical Name or Material 2-(2-Mercapto-4-methyl-5-thiazolyl)ethanol
CAS 1124-01-2
Formula Weight 175.27
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6166803

 
alfa aesar™
H6166803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6166806

 
alfa aesar™
H6166806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61668MD

 
alfa aesar™
H61668MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.27
Assay 95%
Quantity 1g
CAS 1124-01-2
Chemical Name or Material 2-(2-Mercapto-4-methyl-5-thiazolyl)ethanol
Physical Form Solid