missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline], 99%, 98% e.e., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC427520010

 View more versions of this product

Catalog No. AC427520010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Crystalline Powder
294.44
White to light yellow
15, 1297
-118° to -122° (20°C, 589nm) (c=5, in CHCL3)
2, 2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline]
98% ee
99%
MFCD00192243
Authentic
≥98.5%
1g
88° to 91°C
100.0
97.5% min.
C17H30N2O2
131833-93-7
98.5
Glass bottle

Chemical Identifiers

131833-93-7
100.0
C17H30N2O2
MFCD00192243
≥98.5%
98.5
99%
294.44
2, 2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline], 98% ee

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.