Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Hydroxyethylamino)-3-iodopyridine, 95%

$839.03 - $839.03

Chemical Identifiers

Chemical Name or Material 2-(2-Hydroxyethylamino)-3-iodopyridine
CAS 902837-48-3
Molecular Formula C7H9IN2O
Formula Weight 264.07g/mol
MDL Number MFCD07781167
Synonym 2-(3-Iodo-2-pyridylamino)ethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5010406

 
alfa aesar™
H50104-06
5g Each for $839.03
Description & Specifications

Specifications

Quantity 5g
Quantity 5g
Chemical Name or Material 2-(2-Hydroxyethylamino)-3-iodopyridine
CAS 902837-48-3
Assay 95%
Molecular Formula C7H9IN2O
Formula Weight 264.07g/mol
MDL Number MFCD07781167
Synonym 2-(3-Iodo-2-pyridylamino)ethanol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.