Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Hydroxyethyl)pyridine, 99%

$40.99 - $128.38

Chemical Identifiers

Chemical Name or Material 2-(2-Hydroxyethyl)pyridine
CAS 103-74-2
Molecular Formula C7H9NO
Formula Weight 123.16
MDL Number MFCD00006364
Beilstein 111205
Synonym 2-Pyridineethanol; 2-(2-Pyridyl)ethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2357922

  SDS
alfa aesar™
B2357922
100g Each for $40.99

AAB2357936

  SDS
alfa aesar™
B2357936
500g Each for $128.38
Description & Specifications

Specifications

Formula Weight 123.16
CAS 103-74-2
Health Hazard 1 Warning
Flash Point 117°C (243°F)
MDL Number MFCD00006364
Molecular Formula C7H9NO
Synonym 2-Pyridineethanol; 2-(2-Pyridyl)ethanol
Quantity 100g
Assay 99%
RTECS Number UT2970450
Boiling Point 235° to 238°C
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point −8°C
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
TSCA TSCA
Beilstein 111205
Chemical Name or Material 2-(2-Hydroxyethyl)pyridine

Applications
2-(2-Hydroxyethyl)pyridine is a metabolite of the anti-vertigo drug Betahistine. It acts as a reagent for protection of the 5′-phosphate of nucleotides as 2-pyridylethyl (Pet) esters. It is used for carboxyl protection of amino acids. It is also used to cleave such groups as Boc and Fmoc, and also to hydrogenolysis. It was used to prepare mononuclear nickel(II) acetate.

Solubility
Soluble in Chloroform (Slightly), Methanol (Slightly).

Notes
Hygroscopic. Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.