Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Hydroxyethyl)benzeneboronic acid hemiester, 95%

Chemical Identifiers

Chemical Name or Material 2-(2-Hydroxyethyl)benzeneboronic acid hemiester
CAS 19206-51-0
Molecular Formula C8H9BO2
Formula Weight 147.97
MDL Number MFCD08436050
Synonym 3,4-Dihydro-1H-benzo[c][1,2]oxaborinin-1-ol; 2-(2-Hydroxyethyl)benzeneboronic acid dehydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5267203

 
alfa aesar™
H5267203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.97
Chemical Name or Material 2-(2-Hydroxyethyl)benzeneboronic acid hemiester
Assay 95%
Molecular Formula C8H9BO2
MDL Number MFCD08436050
CAS 19206-51-0
Synonym 3,4-Dihydro-1H-benzo[c][1,2]oxaborinin-1-ol; 2-(2-Hydroxyethyl)benzeneboronic acid dehydrate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
2-(2-Hydroxyethyl)benzeneboronic acid hemiester is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.