Please login to your online account to display your discounted pricing

[2-(2-Furyl)phenyl]methanol, ≥97%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC46209CB

 View more versions of this product

Catalog No. CC46209CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 174.2
Quantity 250mg
Assay 97%
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
MDL Number MFCD06203101
CAS 139697-88-4
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Chemical Name or Material [2-(2-Furyl)phenyl]methanol
Packaging Amber glass bottle
Molecular Formula C11H10O2
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material [2-(2-Furyl)phenyl]methanol
CAS 139697-88-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H10O2
Formula Weight 174.2
MDL Number MFCD06203101
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.

SDS & Safety Documents