Please login to your online account to display your discounted pricing

2-(2-Furyl)benzoic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC46201CB

 View more versions of this product

Catalog No. CC46201CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 188.18
Quantity 250mg
Assay 97%
Chemical Name or Material 2-(2-Furyl)benzoic acid
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H8O3
CAS 331942-47-3

Specifications

Chemical Name or Material 2-(2-Furyl)benzoic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H8O3
Formula Weight 188.18