missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2'-Furoin, 98%, Thermo Scientific Chemicals
$157.08 - $1472.73
Chemical Identifiers
CAS | 552-86-3 |
---|---|
Molecular Formula | C10H8O4 |
Molecular Weight (g/mol) | 192.17 |
MDL Number | MFCD00003246 |
InChI Key | MIJRFWVFNKQQDK-UHFFFAOYNA-N |
Synonym | furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone |
PubChem CID | 11100 |
IUPAC Name | 1,2-bis(furan-2-yl)-2-hydroxyethanone |
SMILES | OC(C(=O)C1=CC=CO1)C1=CC=CO1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1527206
|
Thermo Scientific Chemicals
A1527206 |
5 g |
Each for $157.08
|
|
|||||
AAA1527214
|
Thermo Scientific Chemicals
A1527214 |
25 g |
Each for $512.95
|
|
|||||
AAA1527222
|
Thermo Scientific Chemicals
A1527222 |
100 g |
Each for $1,472.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
552-86-3 | |
192.17 | |
MIJRFWVFNKQQDK-UHFFFAOYNA-N | |
11100 | |
OC(C(=O)C1=CC=CO1)C1=CC=CO1 |
C10H8O4 | |
MFCD00003246 | |
furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone | |
1,2-bis(furan-2-yl)-2-hydroxyethanone |
Specifications
552-86-3 | |
C10H8O4 | |
5 g | |
furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone | |
OC(C(=O)C1=CC=CO1)C1=CC=CO1 | |
192.17 | |
192.17 | |
2,2'-Furoin |
137°C to 139°C | |
MFCD00003246 | |
383995 | |
MIJRFWVFNKQQDK-UHFFFAOYNA-N | |
1,2-bis(furan-2-yl)-2-hydroxyethanone | |
11100 | |
98% |
Safety and Handling
EINECSNumber : 209-024-8
RTECSNumber : KM5774095
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only