missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Furoin, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1527222

 View more versions of this product

Catalog No. AAA1527222



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,2'-Furoin
137°C to 139°C
MFCD00003246
383995
MIJRFWVFNKQQDK-UHFFFAOYNA-N
1,2-bis(furan-2-yl)-2-hydroxyethanone
11100
98%
552-86-3
C10H8O4
100 g
furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone
OC(C(=O)C1=CC=CO1)C1=CC=CO1
192.17
192.17

Chemical Identifiers

552-86-3
192.17
MIJRFWVFNKQQDK-UHFFFAOYNA-N
11100
OC(C(=O)C1=CC=CO1)C1=CC=CO1
C10H8O4
MFCD00003246
furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone
1,2-bis(furan-2-yl)-2-hydroxyethanone
Safety and Handling

Safety and Handling

EINECSNumber : 209-024-8

RTECSNumber : KM5774095

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only