Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Furil, 98%

$126.88 - $1,282.26

Chemical Identifiers

Chemical Name or Material 2,2'-Furil
CAS 492-94-4
Molecular Formula C10H6O4
Formula Weight 190.15
MDL Number MFCD00003241
Beilstein 383882
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1538606

  SDS
alfa aesar™
A1538606
5g Each for $126.88

AAA1538614

  SDS
alfa aesar™
A1538614
25g Each for $379.70

AAA1538622

  SDS
alfa aesar™
A1538622
100g Each for $1,282.26
Description & Specifications

Specifications

Formula Weight 190.15
Quantity 5g
Beilstein 383882
CAS 492-94-4
Melting Point 162° to 165°C
Assay 98%
Chemical Name or Material 2,2'-Furil
Molecular Formula C10H6O4
RTECS Number LV0580000
TSCA TSCA
MDL Number MFCD00003241