missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Furil, 98%

Manufacturer:  Alfa Aesar™ A1538622

 View more versions of this product

Catalog No. AAA1538622



Specifications

Specifications

2,2'-Furil
100g
C10H6O4
383882
SXPUVBFQXJHYNS-UHFFFAOYSA-N
1,2-bis(furan-2-yl)ethane-1,2-dione
68119
98%
162°C to 165°C
492-94-4
MFCD00003241
furil, alpha-furil, bipyromucyl, 2,2'-furil, bipryomucyl, di-2-furylglyoxal, difuranylglyoxal, ethanedione, di-2-furanyl, di-2-furanylethanedione, a-furil
C1=COC(=C1)C(=O)C(=O)C2=CC=CO2
190.154
190.15

Chemical Identifiers

492-94-4
190.154
SXPUVBFQXJHYNS-UHFFFAOYSA-N
68119
C1=COC(=C1)C(=O)C(=O)C2=CC=CO2
C10H6O4
MFCD00003241
furil, alpha-furil, bipyromucyl, 2,2'-furil, bipryomucyl, di-2-furylglyoxal, difuranylglyoxal, ethanedione, di-2-furanyl, di-2-furanylethanedione, a-furil
1,2-bis(furan-2-yl)ethane-1,2-dione
Safety and Handling

Safety and Handling

RTECSNumber : LV0580000

TSCA : TSCA

Documents