Learn More
2-(2-Fluorophenyl)pyrrolidine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6169903
Description
2-(2-Fluorophenyl)pyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(2-Fluorophenyl)pyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in freezer(-20°C). Keep container tightly closed. Store in a cool, dry and well-ventilated place.
Specifications
2-(2-Fluorophenyl)pyrrolidine | |
Colorless | |
MFCD02663474 | |
2-2-fluorophenyl pyrrolidine, 2-2-fluorophenyl-pyrrolidine, pyrrolidine,2-2-fluorophenyl, rs-2-2-fluoro-phenyl-pyrrolidine | |
NOJCVVKIBLHAGW-UHFFFAOYSA-N | |
2-(2-fluorophenyl)pyrrolidine | |
3613306 | |
95% |
72216-04-7 | |
C10H12FN | |
1 g | |
Slightly soluble in water. | |
C1CC(NC1)C2=CC=CC=C2F | |
165.211 | |
165.21 | |
Liquid |
Chemical Identifiers
72216-04-7 | |
165.211 | |
NOJCVVKIBLHAGW-UHFFFAOYSA-N | |
3613306 | |
C1CC(NC1)C2=CC=CC=C2F |
C10H12FN | |
MFCD02663474 | |
2-2-fluorophenyl pyrrolidine, 2-2-fluorophenyl-pyrrolidine, pyrrolidine,2-2-fluorophenyl, rs-2-2-fluoro-phenyl-pyrrolidine | |
2-(2-fluorophenyl)pyrrolidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only