Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[2-Fluoro-5-(trifluoromethyl)phenyl]ethanol, 94%

$56.50 - $187.80

Chemical Identifiers

Chemical Name or Material 2-[2-Fluoro-5-(trifluoromethyl)phenyl]ethanol
CAS 886761-79-1
Molecular Formula C9H8F4O
Formula Weight 208.15
MDL Number MFCD06660358
Synonym 2-Fluoro-5-(trifluoromethyl)phenethyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3294303

  SDS
alfa aesar™
H3294303
1g Each for $187.80

AAH32943MD

  SDS
alfa aesar™
H32943MD
250mg Each for $56.50
Description & Specifications

Specifications

Formula Weight 208.15
Chemical Name or Material 2-[2-Fluoro-5-(trifluoromethyl)phenyl]ethanol
Assay 94%
Molecular Formula C9H8F4O
MDL Number MFCD06660358
CAS 886761-79-1
Synonym 2-Fluoro-5-(trifluoromethyl)phenethyl alcohol
Quantity 1g