missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[2-Fluoro-4-(trifluoromethyl)phenyl]ethanol, tech. 90%

$39.57 - $132.07

Chemical Identifiers

CAS 886761-78-0
Molecular Formula C9H8F4O
Formula Weight 208.15
MDL Number MFCD06660357
Synonym 2-Fluoro-4-(trifluoromethyl)phenethyl alcohol
Chemical Name or Material 2-[2-Fluoro-4-(trifluoromethyl)phenyl]ethanol
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3153003 SDS Alfa Aesar™
H3153003
1g Each for $132.07
AAH31530MD SDS Alfa Aesar™
H31530MD
250mg Each for $39.57
Specifications Description & Specifications

Chemical Identifiers

886761-78-0
208.15
2-Fluoro-4-(trifluoromethyl)phenethyl alcohol
Technical Grade
C9H8F4O
MFCD06660357
2-[2-Fluoro-4-(trifluoromethyl)phenyl]ethanol

Specifications

1g
C9H8F4O
MFCD06660357
2-[2-Fluoro-4-(trifluoromethyl)phenyl]ethanol
Technical Grade
886761-78-0
208.15
2-Fluoro-4-(trifluoromethyl)phenethyl alcohol
90%