Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Ethylhexyloxy)-5-methoxy-1,4-benzenedimethanol, 98%

$172.43 - $172.43

Chemical Identifiers

Chemical Name or Material 2-(2-Ethylhexyloxy)-5-methoxy-1,4-benzenedimethanol
CAS 245731-58-2
Molecular Formula C17H28O4
Formula Weight 296.40g/mol
MDL Number MFCD03427231
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5104203

 
alfa aesar™
H51042-03
1g Each for $172.43
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material 2-(2-Ethylhexyloxy)-5-methoxy-1,4-benzenedimethanol
CAS 245731-58-2
Assay 98%
Molecular Formula C17H28O4
Formula Weight 296.40g/mol
MDL Number MFCD03427231