missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2′-Ethylenedianiline, TCI America™

Manufacturer:  TCI America E047225G

 View more versions of this product

Catalog No. E047225G



Specifications

Specifications

2,2′-Ethylenedianiline
74°C
34124-14-6
MFCD00127990
ZYHQGITXIJDDKC-UHFFFAOYSA-N
2-[2-(2-aminophenyl)ethyl]aniline
98690
Crystal-Powder at 20°C
Undesignated
25g
C14H16N2
alpha,alpha′C-Bi-o-toluidine, 2,2′C-Diaminobibenzyl, 2,2′C-Diamino-1,2-diphenylethane
C1=CC=C(C(=C1)CCC2=CC=CC=C2N)N
212.296
212.30
≥98.0% (HPLC,T)

Chemical Identifiers

34124-14-6
212.296
ZYHQGITXIJDDKC-UHFFFAOYSA-N
98690
C1=CC=C(C(=C1)CCC2=CC=CC=C2N)N
C14H16N2
MFCD00127990
alpha,alpha′C-Bi-o-toluidine, 2,2′C-Diaminobibenzyl, 2,2′C-Diamino-1,2-diphenylethane
2-[2-(2-aminophenyl)ethyl]aniline
Safety and Handling

Safety and Handling

TSCA : No

Documents