Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Ethylenebis(1,3-dithiane), 98+%

$39.62 - $122.05

Chemical Identifiers

Chemical Name or Material 2,2'-Ethylenebis(1,3-dithiane)
CAS 14947-53-6
Molecular Formula C10H18S4
Formula Weight 266.51
MDL Number MFCD00040467
Beilstein 1237944
Synonym 1,2-Bis(1,3-dithian-2-yl)ethane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1625503

  SDS
alfa aesar™
L1625503
1g Each for $39.62

AAL1625506

  SDS
alfa aesar™
L1625506
5g Each for $122.05
Description & Specifications

Specifications

Formula Weight 266.51
Chemical Name or Material 2,2'-Ethylenebis(1,3-dithiane)
MDL Number MFCD00040467
CAS 14947-53-6
Molecular Formula C10H18S4
Assay ≥98%
Synonym 1,2-Bis(1,3-dithian-2-yl)ethane
Quantity 1g
Beilstein 1237944
Melting Point 132° to 135°C