Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(2-Ethoxyphenoxy)ethyl bromide, 98%

$52.96 - $169.32

Chemical Identifiers

Chemical Name or Material 2-(2-Ethoxyphenoxy)ethyl bromide
CAS 3259-03-8
Molecular Formula C10H13BrO2
Formula Weight 245.12
MDL Number MFCD02030483
Beilstein 6290077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2030006

 
alfa aesar™
L2030006
5g Each for $52.96

AAL2030014

 
alfa aesar™
L2030014
25g Each for $169.32
Description & Specifications

Specifications

Formula Weight 245.12
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-(2-Ethoxyphenoxy)ethyl bromide
Assay 98%
Molecular Formula C10H13BrO2
MDL Number MFCD02030483
CAS 3259-03-8
Beilstein 6290077
Melting Point 41° to 43°C
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.