missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Dithiosalicylic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 119-80-2
Molecular Formula C14H10O4S2
Molecular Weight (g/mol) 306.34
MDL Number MFCD00002465
InChI Key LBEMXJWGHIEXRA-UHFFFAOYSA-N
Synonym 2,2'-dithiodibenzoic acid, 2,2'-dithiosalicylic acid, benzoic acid, 2,2'-dithiobis, 2-2-carboxyphenyl disulfanyl benzoic acid, bis 2-carboxyphenyl disulfide, bis o-carboxylphenyl disulfide, 2,2-dithiosalicylic acid, 2,2'-dithiobisbenzoic acid, unii-58mgb5279d, 2,2'-dithiodi benzoic acid
PubChem CID 8409
IUPAC Name 2-[(2-carboxyphenyl)disulfanyl]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)SSC2=CC=CC=C2C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC117561000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
117561000
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

119-80-2
306.34
LBEMXJWGHIEXRA-UHFFFAOYSA-N
8409
C1=CC=C(C(=C1)C(=O)O)SSC2=CC=CC=C2C(=O)O
C14H10O4S2
MFCD00002465
2,2'-dithiodibenzoic acid, 2,2'-dithiosalicylic acid, benzoic acid, 2,2'-dithiobis, 2-2-carboxyphenyl disulfanyl benzoic acid, bis 2-carboxyphenyl disulfide, bis o-carboxylphenyl disulfide, 2,2-dithiosalicylic acid, 2,2'-dithiobisbenzoic acid, unii-58mgb5279d, 2,2'-dithiodi benzoic acid
2-[(2-carboxyphenyl)disulfanyl]benzoic acid

Specifications

119-80-2
100.0
98%
C14H10O4S2
MFCD00002465
10, 129
Solubility in water: insoluble. Other solubilities: insoluble in dichloromethane,ether,benzene and,acetone
C1=CC=C(C(=C1)C(=O)O)SSC2=CC=CC=C2C(=O)O
306.34
306.34
2, 2'-Dithiosalicylic acid, 98%
97.5
Authentic
Plastic bottle
S2(C6H4CO2H)2
100g
2,2'-dithiodibenzoic acid, 2,2'-dithiosalicylic acid, benzoic acid, 2,2'-dithiobis, 2-2-carboxyphenyl disulfanyl benzoic acid, bis 2-carboxyphenyl disulfide, bis o-carboxylphenyl disulfide, 2,2-dithiosalicylic acid, 2,2'-dithiobisbenzoic acid, unii-58mgb5279d, 2,2'-dithiodi benzoic acid
LBEMXJWGHIEXRA-UHFFFAOYSA-N
2-[(2-carboxyphenyl)disulfanyl]benzoic acid
8409
98%
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing.

GHS Signal Word: Warning

EINECSNumber : 204-352-8

RTECSNumber : DG9660000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only