Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,2'-Dithiosalicylic acid, 96%

$139.93 - $139.93

Chemical Identifiers

Chemical Name or Material 2,2'-Dithiosalicylic acid
CAS 119-80-2
Molecular Formula C14H10O4S2
Formula Weight 306.36
MDL Number MFCD00002465
Beilstein 2221810
Synonym 2,2'-Dithiodibenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0421336

 
alfa aesar™
L0421336
500g Each for $139.93
Description & Specifications

Specifications

Formula Weight 306.36
Synonym 2,2'-Dithiodibenzoic acid
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2,2'-Dithiosalicylic acid
Molecular Formula C14H10O4S2
MDL Number MFCD00002465
CAS 119-80-2
Beilstein 2221810
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number DG9660000
Odor Odorless
Quantity 500g
TSCA TSCA
Assay 96%
Melting Point 287° to 289°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.