missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Dithiosalicylic acid, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0421336

 View more versions of this product

Catalog No. AAL0421336



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,2'-Dithiosalicylic acid
287°C to 289°C
C14H10O4S2
500 g
2,2'-dithiodibenzoic acid, 2,2'-dithiosalicylic acid, benzoic acid, 2,2'-dithiobis, 2-2-carboxyphenyl disulfanyl benzoic acid, bis 2-carboxyphenyl disulfide, bis o-carboxylphenyl disulfide, 2,2-dithiosalicylic acid, 2,2'-dithiobisbenzoic acid, unii-58mgb5279d, 2,2'-dithiodi benzoic acid
C1=CC=C(C(=C1)C(=O)O)SSC2=CC=CC=C2C(=O)O
306.35
306.36
119-80-2
Odorless
MFCD00002465
2221810
LBEMXJWGHIEXRA-UHFFFAOYSA-N
2-[(2-carboxyphenyl)disulfanyl]benzoic acid
8409
96%

Chemical Identifiers

119-80-2
306.35
LBEMXJWGHIEXRA-UHFFFAOYSA-N
8409
C1=CC=C(C(=C1)C(=O)O)SSC2=CC=CC=C2C(=O)O
C14H10O4S2
MFCD00002465
2,2'-dithiodibenzoic acid, 2,2'-dithiosalicylic acid, benzoic acid, 2,2'-dithiobis, 2-2-carboxyphenyl disulfanyl benzoic acid, bis 2-carboxyphenyl disulfide, bis o-carboxylphenyl disulfide, 2,2-dithiosalicylic acid, 2,2'-dithiobisbenzoic acid, unii-58mgb5279d, 2,2'-dithiodi benzoic acid
2-[(2-carboxyphenyl)disulfanyl]benzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

EINECSNumber : 204-352-8

RTECSNumber : DG9660000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only