missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dithiodipyridine, 98%

$204.83 - $2360.15

Chemical Identifiers


Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC162230050
SDS View Documents
Thermo Scientific Chemicals
162230050
5 g Glass bottle
Each for $204.83
Only null left
 
AC162230250
SDS View Documents
Thermo Scientific Chemicals
162230250
25 g Glass bottle
Each for $659.56
Only null left
 
AC162231000
SDS View Documents
Thermo Scientific Chemicals
162231000
100 g Glass bottle
Each for $2,360.15
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

0.0

Specifications

56°C to 60°C
Colorless to Tan
100.0
5 g
C10H8N2S2
MFCD00006287
06,246; 13,156
2, 2';-Dipyridyl disulfide
HAXFWIACAGNFHA-UHFFFAOYSA-N
9-[(3aS,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-3H-purin-6-one
129317592
98%
Glass bottle
2,2′-Dithiodipyridine
2127-03-9
97.5
Liquid
97.5% min. (GC)
(C5H4N)S-S(C5H4N)
21, 45
210°C
Solubility in water: 5g/L (20°C). Other solubilities: slightly soluble in methanol,acetone,toluene and,isopropanol,practically insoluble in heptane and cyclohexane
S(SC1=CC=CC=N1)C1=CC=CC=N1
220.32
220.32
Authentic
(5% soln. in methanol) clear
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 218-343-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only