missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Dithiodipyridine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC162231000

 View more versions of this product

Catalog No. AC162231000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5% (GC)
Liquid
Clear colorless to yellow to tan
21, 45
100.0
2, 2';-Dithiodipyridine
2127-03-9
2, 2';-Dipyridyl disulfide
Glass bottle
(C5H4N)S-S(C5H4N)
220.32
100g
55° to 60°C
06,246; 13,156
MFCD00006287
98%
97.5
Authentic
(5% soln. in methanol) clear
C10H8N2S2

Chemical Identifiers

2127-03-9
100.0
C10H8N2S2
220.32
2, 2';-Dithiodipyridine
97.5
98%
(C5H4N)S-S(C5H4N)
MFCD00006287
≥97.5% (GC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning