Learn More
2,2'-Dithiodipyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 162230050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,2′-Dithiodipyridine | |
2127-03-9 | |
97.5 | |
Liquid | |
97.5% min. (GC) | |
(C5H4N)S-S(C5H4N) | |
21, 45 | |
210°C | |
Solubility in water: 5g/L (20°C). Other solubilities: slightly soluble in methanol,acetone,toluene and,isopropanol,practically insoluble in heptane and cyclohexane | |
S(SC1=CC=CC=N1)C1=CC=CC=N1 | |
220.32 | |
220.32 | |
Authentic | |
(5% soln. in methanol) clear |
56°C to 60°C | |
Colorless to Tan | |
100.0 | |
5 g | |
C10H8N2S2 | |
MFCD00006287 | |
06,246; 13,156 | |
2, 2';-Dipyridyl disulfide | |
HAXFWIACAGNFHA-UHFFFAOYSA-N | |
9-[(3aS,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-3H-purin-6-one | |
129317592 | |
98% | |
Glass bottle |
Chemical Identifiers
2127-03-9 | |
220.32 | |
HAXFWIACAGNFHA-UHFFFAOYSA-N | |
129317592 | |
S(SC1=CC=CC=N1)C1=CC=CC=N1 |
C10H8N2S2 | |
MFCD00006287 | |
2, 2';-Dipyridyl disulfide | |
9-[(3aS,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-3H-purin-6-one |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Warning
EINECSNumber : 218-343-1
RUO – Research Use Only