missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Dithiodipyridine, 98%, ACROS Organics™

Manufacturer:  Acros Organics 162230250

 View more versions of this product

Catalog No. AC162230250



Specifications

Specifications

(5% soln. in methanol) clear
Authentic
Colorless to Tan
25g
97.5
98%
(C5H4N)S-S(C5H4N)
21, 45
2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide
C1=CC=NC(=C1)SSC2=CC=CC=N2
220.31
220.32
≥97.5% (GC)
2, 2';-Dithiodipyridine
Glass bottle
55°C to 60°C
2127-03-9
100.0
C10H8N2S2
MFCD00006287
06,246; 13,156
HAXFWIACAGNFHA-UHFFFAOYSA-N
2-(pyridin-2-yldisulfanyl)pyridine
65093
Liquid

Chemical Identifiers

2127-03-9
220.31
HAXFWIACAGNFHA-UHFFFAOYSA-N
65093
C1=CC=NC(=C1)SSC2=CC=CC=N2
C10H8N2S2
MFCD00006287
2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide
2-(pyridin-2-yldisulfanyl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents