Learn More
2,2'-Dipyridyl disulfide, 98%, Thermo Scientific Chemicals
$129.44 - $1,273.26
Chemical Identifiers
CAS | 2127-03-9 |
---|---|
Molecular Formula | C10H8N2S2 |
Molecular Weight (g/mol) | 220.31 |
MDL Number | MFCD00006287 |
InChI Key | HAXFWIACAGNFHA-UHFFFAOYSA-N |
Synonym | 2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide |
PubChem CID | 65093 |
IUPAC Name | 2-(pyridin-2-yldisulfanyl)pyridine |
SMILES | C1=CC=NC(=C1)SSC2=CC=CC=N2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1111806
|
Thermo Scientific Chemicals
A1111806 |
5 g |
N/A
|
|
|||||
AAA1111814
|
Thermo Scientific Chemicals
A1111814 |
25 g |
N/A
|
|
|||||
AAA1111822
|
Thermo Scientific Chemicals
A1111822 |
100 g |
N/A
|
|
|||||
Description
2,2'-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2′-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.
Solubility
Soluble in water, benzene, tetrahydrofuran, dichloromethane, acetonitrile and dimethyformamide.
Notes
Incompatible with oxidizing agents and metals.
Chemical Identifiers
2127-03-9 | |
220.31 | |
HAXFWIACAGNFHA-UHFFFAOYSA-N | |
65093 | |
C1=CC=NC(=C1)SSC2=CC=CC=N2 |
C10H8N2S2 | |
MFCD00006287 | |
2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide | |
2-(pyridin-2-yldisulfanyl)pyridine |
Specifications
2127-03-9 | |
210°C (410°F) | |
C10H8N2S2 | |
5 g | |
2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide | |
HAXFWIACAGNFHA-UHFFFAOYSA-N | |
2-(pyridin-2-yldisulfanyl)pyridine | |
65093 | |
98% |
56°C to 59°C | |
98% | |
MFCD00006287 | |
154629 | |
Soluble in water,benzene,tetrahydrofuran,dichloromethane,acetonitrile and dimethyformamide. | |
C1=CC=NC(=C1)SSC2=CC=CC=N2 | |
220.31 | |
220.32 | |
2, 2′-Dipyridyl disulfide |
Safety and Handling
GHS H Statement
H315-H319-H335
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 218-343-1
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only