missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Dipyridyl disulfide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1111806

 View more versions of this product

Catalog No. AAA1111806



Description

Description

2,2'-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.

Solubility
Soluble in water, benzene, tetrahydrofuran, dichloromethane, acetonitrile and dimethyformamide.

Notes
Incompatible with oxidizing agents and metals.
Specifications

Specifications

2, 2′-Dipyridyl disulfide
56°C to 59°C
98%
MFCD00006287
154629
Soluble in water,benzene,tetrahydrofuran,dichloromethane,acetonitrile and dimethyformamide.
C1=CC=NC(=C1)SSC2=CC=CC=N2
220.31
220.32
2127-03-9
210°C (410°F)
C10H8N2S2
5 g
2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide
HAXFWIACAGNFHA-UHFFFAOYSA-N
2-(pyridin-2-yldisulfanyl)pyridine
65093
98%

Chemical Identifiers

2127-03-9
220.31
HAXFWIACAGNFHA-UHFFFAOYSA-N
65093
C1=CC=NC(=C1)SSC2=CC=CC=N2
C10H8N2S2
MFCD00006287
2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide
2-(pyridin-2-yldisulfanyl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 218-343-1

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only