Learn More
2,2'-Dipyridyl, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 117500250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
2, 2'-Dipyridyl | |
99.0 | |
69.0°C to 72.0°C | |
272.0°C to 273.0°C | |
Authentic | |
Glass bottle | |
MFCD00006212 | |
23,199 | |
15,1241 | |
Solubility in water: slightly soluble (5.5g/L 22°C). Other solubilities: very soluble in ethanol (500g/L 25°C), ether,, benzene, chloroform and petroleum ether | |
C1=CC=C(N=C1)C1=CC=CC=N1 | |
156.19 | |
1.51 mm2/s (90°C) | |
156.19 | |
Crystalline Powder |
366-18-7 | |
100.0 | |
White | |
121°C | |
99% min. (GC) | |
C10H8N2 | |
25 g | |
16,25 | |
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl | |
ROFVEXUMMXZLPA-UHFFFAOYSA-N | |
2-pyridin-2-ylpyridine | |
1474 | |
CHEBI:30351 | |
99+% |
Chemical Identifiers
366-18-7 | |
156.19 | |
ROFVEXUMMXZLPA-UHFFFAOYSA-N | |
1474 | |
2-pyridin-2-ylpyridine |
C10H8N2 | |
MFCD00006212 | |
2,2'-bipyridine, 2,2'-dipyridyl, 2,2'-bipyridyl, bipyridine, 2,2'-dipyridine, 2-2-pyridyl pyridine, 2,2'-bipyridin, alpha,alpha'-dipyridyl, alpha,alpha'-bipyridyl, 2,2-bipyridyl | |
CHEBI:30351 | |
C1=CC=C(N=C1)C1=CC=CC=N1 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or d
GHS Signal Word: Danger
EINECSNumber : 206-674-4
RUO – Research Use Only