missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Diphenylpropionitrile, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1799109

 View more versions of this product

Catalog No. AAA1799109



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,2-Diphenylpropionitrile
1.108
>110°C (230°F)
1.572
10 g
1955952
DPVHBXFSKLKYIQ-UHFFFAOYSA-N
2,2-diphenylpropanenitrile
79677
97%
5558-67-8
144°C to 145°C (2 mmHg)
C15H13N
MFCD00001846
UN3276
2,2-diphenylpropionitrile, 1,1-diphenylethyl cyanide, 2,2-diphenylpropiononitrile, 2,2-diphenyl propionitrile, .alpha.,.alpha.-diphenylpropionitrile, pubchem12547, acmc-209lnw, 2,2-diphenylpropanitrile, 2,2-diphenyl-propionitrile, 2, 2-diphenyl propionitrile
CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2
207.276
207.28

Chemical Identifiers

5558-67-8
207.276
DPVHBXFSKLKYIQ-UHFFFAOYSA-N
2,2-diphenylpropanenitrile
C15H13N
MFCD00001846
79677
CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 226-925-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only