Please login to your online account to display your discounted pricing

2,2-Diphenylethylamine 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 2-Diphenylethylamine
Name Note 96%
CAS 3963-62-0
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C14H15N
Linear Formula (C6H5)2CHCH2NH2
Formula Weight 197.28
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC117250050

  SDS
thermo fisher scientific
117250050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC117250250

  SDS
thermo fisher scientific
117250250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Formula Weight 197.28
Physical Form Fused Solid
Quantity 5g
Color White to light yellow
Beilstein 12, 1327
Melting Point 43° to 49°C
EINECS Number 223-565-7
TSCA TSCA
CAS Max % 100.0
CAS Min % 95.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 2, 2-Diphenylethylamine
CAS 3963-62-0
MDL Number MFCD00008143
Assay Percent Range 96%
Name Note 96%
Molecular Formula C14H15N
Linear Formula (C6H5)2CHCH2NH2
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.