missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Diphenyl-1-picrylhydrazyl (free radical), 95%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 44150MD

 View more versions of this product

Catalog No. AA44150MD



Specifications

Specifications

2,2-Diphenyl-1-picrylhydrazyl (free radical)
250mg
C18H12N5O6
14,3330
Soluble in dimethyl formamide,ethanol,chloroform,ether,carbon disulfinde,acetone,oils,carbon tetrachloride,glacial acetic acid and toluene. Insoluble in water.
Powder
Black to Green
1898-66-4
MFCD00007231
DPPH
394.32
95%

Chemical Identifiers

1898-66-4
C18H12N5O6
MFCD00007231
DPPH
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

EINECSNumber : 217-591-8

RTECSNumber : MW3250000

TSCA : Yes

Recommended Storage : Keep cold

Documents